The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002949.3 | NP_002940.2 | 1 - 2 | 204 | 113 |
GenBank mRNA | BC002344.2 | - | 1 - 2 | 189 | 113 |
BC007497.2 | - | 1 - 2 | 86 | 113 | |
DA648796.1 | - | 1 - 2 | 204 | 113 |