The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184970.1 | NP_001171899.1 | 1 - 2 | 153 | 156 |
XM_005261319.3 | XP_005261376.1 | 1 - 2 | 665 | 156 | |
XM_011529846.1 | XP_011528148.1 | 1 - 2 | 665 | 156 | |
XM_011529853.1 | XP_011528155.1 | 1 - 2 | 665 | 156 | |
GenBank mRNA | AF128536.1 | - | 1 - 2 | 126 | 156 |