Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
Chromosome Location
Chr.1: 241593150 - 241640399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC051900.1 - 1 - 2 390 111
BC073872.1 - 1 - 2 441 111

View Full Product Details