The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031283.2 | NP_112573.1 | 4 - 5 | 744 | 74 |
XM_006712109.2 | XP_006712172.1 | 4 - 5 | 534 | 74 | |
GenBank mRNA | AB031046.1 | - | 4 - 5 | 593 | 74 |
BC058894.1 | - | 4 - 5 | 753 | 74 |