The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002318.2 | NP_002309.1 | 13 - 14 | 2587 | 80 |
GenBank mRNA | AK124508.1 | - | 3 - 4 | 1730 | 80 |
AK222477.1 | - | 13 - 14 | 2498 | 80 | |
AK312266.1 | - | 13 - 14 | 2499 | 80 | |
BC000594.2 | - | 13 - 14 | 2457 | 80 | |
KF975332.1 | - | 12 - 13 | 2188 | 80 | |
U89942.1 | - | 13 - 14 | 2495 | 80 |