Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL5
Entrez Gene ID
83416
Gene Name
Fc receptor like 5
Gene Aliases
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
Chromosome Location
Chr.1: 157513377 - 157552520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195388.1 NP_001182317.1 6 - 7 1283 69
NM_031281.2 NP_112571.2 6 - 7 1283 69
XM_011510030.1 XP_011508332.1 5 - 6 985 69
XM_011510031.1 XP_011508333.1 6 - 7 1281 69
XM_011510033.1 XP_011508335.1 6 - 7 1281 69
GenBank mRNA AF343662.1 - 6 - 7 1218 69
AF343663.1 - 6 - 7 1218 69
AF343664.1 - 6 - 7 1218 69
AF369794.2 - 6 - 7 1283 69
AF397453.1 - 6 - 7 1125 69
AK300010.1 - 5 - 6 1009 69
AK301356.1 - 7 - 8 1323 69
BC101066.2 - 6 - 7 1153 69
BC101067.1 - 6 - 7 1153 69
BC101069.2 - 6 - 7 1153 69

View Full Product Details