The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278599.1 | NP_001265528.1 | 15 - 16 | 1771 | 72 |
NM_001699.5 | NP_001690.2 | 17 - 18 | 2366 | 72 | |
NM_021913.4 | NP_068713.2 | 18 - 19 | 2393 | 72 | |
GenBank mRNA | AK309139.1 | - | 15 - 16 | 1771 | 72 |
BC032229.1 | - | 18 - 19 | 2364 | 72 | |
M76125.1 | - | 17 - 18 | 2330 | 72 | |
S65125.1 | - | 18 - 19 | 2234 | 72 | |
X57019.1 | - | 18 - 19 | 2234 | 72 |