The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278599.1 | NP_001265528.1 | 6 - 7 | 859 | 131 |
NM_021913.4 | NP_068713.2 | 9 - 10 | 1481 | 131 | |
GenBank mRNA | AK309139.1 | - | 6 - 7 | 859 | 131 |
AK310881.1 | - | 9 - 10 | 1302 | 131 | |
BC032229.1 | - | 9 - 10 | 1452 | 131 |