Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGEF2
Entrez Gene ID
9181
Gene Name
Rho/Rac guanine nucleotide exchange factor 2
Gene Aliases
GEF, GEF-H1, GEFH1, LFP40, P40
Chromosome Location
Chr.1: 155946839 - 155991263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162383.1 NP_001155855.1 15 - 16 2102 91
NM_001162384.1 NP_001155856.1 15 - 16 2099 91
NM_004723.3 NP_004714.2 15 - 16 2027 91
XM_005245587.2 XP_005245644.1 15 - 16 2155 91
XM_005245588.2 XP_005245645.1 15 - 16 2152 91
XM_005245589.4 XP_005245646.1 15 - 16 2308 91
XM_005245590.3 XP_005245647.1 15 - 16 2030 91
XM_005245591.3 XP_005245648.1 16 - 17 2146 91
XM_005245592.2 XP_005245649.1 15 - 16 2299 91
XM_005245593.4 XP_005245650.1 15 - 16 2030 91
XM_005245594.2 XP_005245651.1 15 - 16 3016 91
XM_006711622.3 XP_006711685.1 15 - 16 3293 91
XM_006711623.3 XP_006711686.1 14 - 15 2029 91
XM_006711624.3 XP_006711687.1 14 - 15 2056 91
XM_006711625.3 XP_006711688.1 14 - 15 1962 91
XM_006711626.2 XP_006711689.1 14 - 15 2948 91
XM_011510136.2 XP_011508438.2 19 - 20 2711 91
XM_011510137.2 XP_011508439.2 19 - 20 2714 91
XM_011510139.2 XP_011508441.1 19 - 20 2714 91
XM_011510141.2 XP_011508443.1 15 - 16 2832 91
XM_011510143.2 XP_011508445.1 11 - 12 1578 91
XM_017002794.1 XP_016858283.1 19 - 20 2711 91
XM_017002795.1 XP_016858284.1 15 - 16 2046 91
XM_017002796.1 XP_016858285.1 15 - 16 3290 91
XM_017002797.1 XP_016858286.1 15 - 16 2030 91
XM_017002798.1 XP_016858287.1 14 - 15 3225 91
XM_017002799.1 XP_016858288.1 14 - 15 2026 91
GenBank mRNA AB014551.3 - 19 - 20 2370 91
AF486838.1 - 15 - 16 1981 91
AL512715.1 - 15 - 16 2012 91
BC020567.1 - 15 - 16 2084 91
BT007407.1 - 14 - 15 1903 91
U72206.1 - 15 - 16 2086 91
X15610.1 - 1 - 2 94 91

View Full Product Details