The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054027.4 | NP_473368.1 | 5 - 6 | 1015 | 67 |
XM_011514067.1 | XP_011512369.1 | 5 - 6 | 978 | 67 | |
XM_017009644.1 | XP_016865133.1 | 5 - 6 | 615 | 67 | |
GenBank mRNA | AB046801.1 | - | 5 - 6 | 845 | 67 |
AF274753.1 | - | 5 - 6 | 752 | 67 | |
AK001799.1 | - | 1 - 2 | 140 | 67 | |
AK222631.1 | - | - | 713 | 67 | |
AK315012.1 | - | 5 - 6 | 713 | 67 | |
AY358503.1 | - | 5 - 6 | 766 | 67 | |
BC009835.2 | - | 5 - 6 | 940 | 67 | |
BC014526.2 | - | 5 - 6 | 740 | 67 |