The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054027.4 | NP_473368.1 | 6 - 7 | 1155 | 112 |
XM_011514067.1 | XP_011512369.1 | 6 - 7 | 1118 | 112 | |
XM_017009644.1 | XP_016865133.1 | 6 - 7 | 755 | 112 | |
GenBank mRNA | AB046801.1 | - | 6 - 7 | 985 | 112 |
AK001799.1 | - | 2 - 3 | 280 | 112 | |
AK130676.1 | - | 1 - 2 | 119 | 112 | |
AK222631.1 | - | - | 853 | 112 | |
AK315012.1 | - | 6 - 7 | 853 | 112 | |
AY358503.1 | - | 6 - 7 | 906 | 112 | |
BC009835.2 | - | 6 - 7 | 1080 | 112 | |
BC014526.2 | - | 6 - 7 | 880 | 112 |