The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317062.1 | NP_001303991.1 | 4 - 5 | 635 | 63 |
NM_001317063.1 | NP_001303992.1 | 4 - 5 | 635 | 63 | |
NM_024102.3 | NP_077007.1 | 4 - 5 | 635 | 63 | |
NR_133654.1 | 3 - 4 | 493 | 63 | ||
GenBank mRNA | AB073603.1 | - | 4 - 5 | 583 | 63 |
AF052183.1 | - | 4 - 5 | 521 | 63 | |
AF478464.1 | - | 4 - 5 | 496 | 63 | |
AK022860.1 | - | 4 - 5 | 565 | 63 | |
AK223189.1 | - | 4 - 5 | 565 | 63 | |
AK296163.1 | - | 3 - 4 | 437 | 63 | |
AL557974.3 | - | 4 - 5 | 551 | 63 | |
AY225316.1 | - | 4 - 5 | 521 | 63 | |
BC001679.2 | - | 4 - 5 | 556 | 63 | |
BC006477.1 | - | 4 - 5 | 556 | 63 | |
BC009411.1 | - | 4 - 5 | 568 | 63 | |
BC011778.2 | - | 4 - 5 | 575 | 63 | |
BC016946.1 | - | 4 - 5 | 535 | 63 | |
BI195029.1 | - | 3 - 4 | 307 | 63 |