Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTPN23
Entrez Gene ID
25930
Gene Name
protein tyrosine phosphatase, non-receptor type 23
Gene Aliases
HD-PTP, HDPTP, PTP-TD14
Chromosome Location
Chr.3: 47380982 - 47413441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304482.1 NP_001291411.1 7 - 8 743 77
NM_015466.3 NP_056281.1 8 - 9 871 77
XM_005265031.2 XP_005265088.2 8 - 9 822 77
GenBank mRNA AB025194.1 - 8 - 9 819 77
AB040904.2 - 8 - 9 871 77
AF290614.1 - 8 - 9 817 77
AK055157.1 - 8 - 9 842 77
AK289502.1 - 8 - 9 820 77
AK295877.1 - 8 - 9 791 77
AK295888.1 - 7 - 8 734 77
AK299906.1 - 7 - 8 714 77
BC089042.1 - 8 - 9 789 77

View Full Product Details