Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGM4
Entrez Gene ID
7047
Gene Name
transglutaminase 4
Gene Aliases
TGP, hTGP
Chromosome Location
Chr.3: 44874592 - 44914601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003241.3 NP_003232.2 11 - 12 1669 75
XM_011534042.1 XP_011532344.1 12 - 13 1818 75
GenBank mRNA AK300523.1 - 11 - 12 1538 75
AK312936.1 - 11 - 12 1667 75
BC007003.1 - 11 - 12 1664 75
L34840.1 - 11 - 12 1640 75
U31905.1 - 11 - 12 1640 75

View Full Product Details