Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGM4
Entrez Gene ID
7047
Gene Name
transglutaminase 4
Gene Aliases
TGP, hTGP
Chromosome Location
Chr.3: 44874592 - 44914601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003241.3 NP_003232.2 4 - 5 496 102
XM_011534042.1 XP_011532344.1 5 - 6 645 102
GenBank mRNA AK300523.1 - 4 - 5 365 102
AK312936.1 - 4 - 5 494 102
BC007003.1 - 4 - 5 491 102
L34840.1 - 4 - 5 467 102
U31905.1 - 4 - 5 467 102
U79008.1 - 3 - 4 360 102
Y09559.1 - 3 - 4 360 102

View Full Product Details