The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136479.2 | NP_001129951.1 | 2 - 3 | 256 | 73 |
NM_001280559.1 | NP_001267488.1 | 2 - 3 | 580 | 73 | |
NM_001280560.1 | NP_001267489.1 | 2 - 3 | 580 | 73 | |
NM_030920.4 | NP_112182.1 | 2 - 3 | 580 | 73 | |
XM_005245513.3 | XP_005245570.1 | 2 - 3 | 578 | 73 | |
XM_005245514.3 | XP_005245571.1 | 2 - 3 | 578 | 73 | |
XM_017002418.1 | XP_016857907.1 | 2 - 3 | 578 | 73 | |
GenBank mRNA | AK092672.1 | - | 2 - 3 | 541 | 73 |
AK095228.1 | - | 2 - 3 | 256 | 73 | |
AK223469.1 | - | 2 - 3 | 447 | 73 | |
AK303333.1 | - | 2 - 3 | 380 | 73 | |
AL832664.1 | - | 2 - 3 | 446 | 73 | |
AL832674.1 | - | 2 - 3 | 582 | 73 | |
AL833660.1 | - | 2 - 3 | 334 | 73 | |
AY057381.1 | - | 2 - 3 | 519 | 73 | |
BC003380.1 | - | 2 - 3 | 399 | 73 |