The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159508.1 | NP_001152980.1 | 3 - 4 | 711 | 62 |
NM_002225.3 | NP_002216.2 | 4 - 5 | 801 | 62 | |
XM_005254350.2 | XP_005254407.1 | 4 - 5 | 467 | 62 | |
XM_006720492.2 | XP_006720555.1 | 4 - 5 | 467 | 62 | |
XM_006720495.2 | XP_006720558.1 | 4 - 5 | 467 | 62 | |
XM_017022149.1 | XP_016877638.1 | 4 - 5 | 586 | 62 | |
XM_017022150.1 | XP_016877639.1 | 4 - 5 | 585 | 62 | |
XM_017022151.1 | XP_016877640.1 | 4 - 5 | 585 | 62 | |
XM_017022152.1 | XP_016877641.1 | 4 - 5 | 582 | 62 | |
XM_017022153.1 | XP_016877642.1 | 4 - 5 | 584 | 62 | |
XM_017022154.1 | XP_016877643.1 | 4 - 5 | 723 | 62 | |
XM_017022155.1 | XP_016877644.1 | 4 - 5 | 583 | 62 | |
XM_017022156.1 | XP_016877645.1 | 4 - 5 | 868 | 62 | |
XM_017022157.1 | XP_016877646.1 | 4 - 5 | 585 | 62 | |
XM_017022158.1 | XP_016877647.1 | 4 - 5 | 585 | 62 | |
GenBank mRNA | AK122922.1 | - | 3 - 4 | 401 | 62 |
AK315296.1 | - | 4 - 5 | 467 | 62 | |
BC017202.2 | - | 4 - 5 | 469 | 62 | |
BT007145.1 | - | 4 - 5 | 458 | 62 | |
KU178150.1 | - | 4 - 5 | 458 | 62 | |
KU178151.1 | - | 3 - 4 | 368 | 62 | |
M34192.1 | - | 4 - 5 | 473 | 62 |