The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 12 - 13 | 2059 | 77 |
NM_001323615.1 | NP_001310544.1 | 11 - 12 | 1921 | 77 | |
NM_001323616.1 | NP_001310545.1 | 13 - 14 | 2191 | 77 | |
NM_001323617.1 | NP_001310546.1 | 13 - 14 | 2026 | 77 | |
NM_015327.2 | NP_056142.2 | 12 - 13 | 1998 | 77 | |
XM_017000842.1 | XP_016856331.1 | 8 - 9 | 1944 | 77 | |
XM_017000843.1 | XP_016856332.1 | 11 - 12 | 1926 | 77 | |
GenBank mRNA | AB029012.2 | - | 12 - 13 | 2000 | 77 |
AB085691.1 | - | 12 - 13 | 1926 | 77 | |
AL137738.1 | - | 1 - 2 | 100 | 77 | |
AY168922.1 | - | 12 - 13 | 1852 | 77 | |
AY336728.1 | - | 12 - 13 | 1849 | 77 | |
BC007453.2 | - | 1 - 2 | 445 | 77 | |
BC038296.1 | - | 12 - 13 | 1999 | 77 |