Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PARP8
- Entrez Gene ID
-
79668
- Gene Name
- poly(ADP-ribose) polymerase family member 8
- Gene Aliases
-
ARTD16, pART16
- Chromosome Location
-
Chr.5: 50665899 - 50846522 on Build GRCh38
View Full Product Details