Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSWIM2
Entrez Gene ID
151112
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
MEX, ZZZ2
Chromosome Location
Chr.2: 186827480 - 186849170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182521.2 NP_872327.2 3 - 4 320 103
XM_011510712.1 XP_011509014.1 3 - 4 320 103
GenBank mRNA AK128006.1 - 3 - 4 320 103
BC031094.1 - 3 - 4 319 103

View Full Product Details