Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 15 - 16 2068 101
NM_020843.2 NP_065894.2 14 - 15 1925 101
XM_005254417.2 XP_005254474.1 15 - 16 2100 101
XM_005254419.2 XP_005254476.1 15 - 16 2082 101
XM_011521650.1 XP_011519952.1 16 - 17 2185 101
XM_011521652.2 XP_011519954.1 14 - 15 1896 101
XM_011521653.2 XP_011519955.1 14 - 15 1980 101
XM_011521654.2 XP_011519956.1 8 - 9 1244 101
XM_017022266.1 XP_016877755.1 15 - 16 2105 101
XM_017022267.1 XP_016877756.1 15 - 16 2087 101
XM_017022268.1 XP_016877757.1 15 - 16 2048 101
XM_017022269.1 XP_016877758.1 15 - 16 2080 101
XM_017022270.1 XP_016877759.1 16 - 17 2238 101
XM_017022271.1 XP_016877760.1 15 - 16 2106 101
XM_017022272.1 XP_016877761.1 15 - 16 2100 101
XM_017022273.1 XP_016877762.1 14 - 15 1962 101
XM_017022274.1 XP_016877763.1 15 - 16 2105 101
XM_017022275.1 XP_016877764.1 14 - 15 1937 101
XM_017022276.1 XP_016877765.1 12 - 13 1792 101
XM_017022277.1 XP_016877766.1 14 - 15 1919 101
XM_017022278.1 XP_016877767.1 13 - 14 1801 101
XM_017022279.1 XP_016877768.1 15 - 16 2102 101
XM_017022280.1 XP_016877769.1 15 - 16 2102 101
XM_017022281.1 XP_016877770.1 15 - 16 2102 101
XM_017022282.1 XP_016877771.1 15 - 16 2101 101
XM_017022283.1 XP_016877772.1 15 - 16 2082 101
GenBank mRNA AB040887.1 - 14 - 15 1930 101
AF119814.1 - 8 - 9 1132 101
AF242528.1 - 14 - 15 1900 101
AK056095.1 - 15 - 16 2089 101
AK056515.1 - 17 - 18 2209 101
BX647285.1 - 15 - 16 2068 101

View Full Product Details