Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 20 - 21 2717 79
NM_020843.2 NP_065894.2 19 - 20 2574 79
XM_005254417.2 XP_005254474.1 20 - 21 2749 79
XM_005254419.2 XP_005254476.1 20 - 21 2731 79
XM_011521650.1 XP_011519952.1 21 - 22 2834 79
XM_011521652.2 XP_011519954.1 19 - 20 2545 79
XM_011521653.2 XP_011519955.1 19 - 20 2629 79
XM_011521654.2 XP_011519956.1 13 - 14 1893 79
XM_011521656.2 XP_011519958.1 6 - 7 750 79
XM_017022266.1 XP_016877755.1 20 - 21 2754 79
XM_017022267.1 XP_016877756.1 20 - 21 2736 79
XM_017022268.1 XP_016877757.1 20 - 21 2697 79
XM_017022269.1 XP_016877758.1 20 - 21 2729 79
XM_017022270.1 XP_016877759.1 21 - 22 2887 79
XM_017022271.1 XP_016877760.1 20 - 21 2755 79
XM_017022272.1 XP_016877761.1 20 - 21 2749 79
XM_017022273.1 XP_016877762.1 19 - 20 2611 79
XM_017022274.1 XP_016877763.1 20 - 21 2754 79
XM_017022275.1 XP_016877764.1 19 - 20 2586 79
XM_017022276.1 XP_016877765.1 17 - 18 2441 79
XM_017022277.1 XP_016877766.1 19 - 20 2568 79
XM_017022278.1 XP_016877767.1 18 - 19 2450 79
XM_017022279.1 XP_016877768.1 20 - 21 2751 79
XM_017022280.1 XP_016877769.1 20 - 21 2751 79
XM_017022281.1 XP_016877770.1 20 - 21 2751 79
GenBank mRNA AB040887.1 - 19 - 20 2579 79
AF119814.1 - 13 - 14 1781 79
AF242528.1 - 19 - 20 2549 79
AK025663.1 - 4 - 5 452 79
AK056515.1 - 22 - 23 2858 79

View Full Product Details