Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 24 - 25 3156 80
NM_020843.2 NP_065894.2 23 - 24 3013 80
XM_005254417.2 XP_005254474.1 24 - 25 3188 80
XM_005254419.2 XP_005254476.1 24 - 25 3170 80
XM_011521650.1 XP_011519952.1 25 - 26 3273 80
XM_011521652.2 XP_011519954.1 23 - 24 2984 80
XM_011521653.2 XP_011519955.1 23 - 24 3068 80
XM_011521654.2 XP_011519956.1 17 - 18 2332 80
XM_011521656.2 XP_011519958.1 10 - 11 1189 80
XM_017022266.1 XP_016877755.1 24 - 25 3193 80
XM_017022267.1 XP_016877756.1 24 - 25 3175 80
XM_017022268.1 XP_016877757.1 24 - 25 3136 80
XM_017022269.1 XP_016877758.1 24 - 25 3168 80
XM_017022270.1 XP_016877759.1 25 - 26 3326 80
XM_017022271.1 XP_016877760.1 24 - 25 3194 80
XM_017022272.1 XP_016877761.1 24 - 25 3188 80
XM_017022273.1 XP_016877762.1 23 - 24 3050 80
XM_017022274.1 XP_016877763.1 24 - 25 3193 80
XM_017022275.1 XP_016877764.1 23 - 24 3025 80
XM_017022276.1 XP_016877765.1 21 - 22 2880 80
XM_017022277.1 XP_016877766.1 23 - 24 3007 80
XM_017022278.1 XP_016877767.1 22 - 23 2889 80
XM_017022279.1 XP_016877768.1 24 - 25 3190 80
XM_017022280.1 XP_016877769.1 24 - 25 3190 80
XM_017022281.1 XP_016877770.1 24 - 25 3190 80
GenBank mRNA AB040887.1 - 23 - 24 3018 80
AF119814.1 - 17 - 18 2220 80
AF242528.1 - 23 - 24 2988 80
AK025663.1 - 8 - 9 891 80
BX647285.1 - 25 - 26 3156 80

View Full Product Details