Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 6 - 7 655 61
NM_020843.2 NP_065894.2 6 - 7 671 61
XM_005254417.2 XP_005254474.1 7 - 8 828 61
XM_005254419.2 XP_005254476.1 7 - 8 828 61
XM_011521650.1 XP_011519952.1 8 - 9 913 61
XM_011521652.2 XP_011519954.1 6 - 7 624 61
XM_011521653.2 XP_011519955.1 6 - 7 708 61
XM_017022266.1 XP_016877755.1 7 - 8 833 61
XM_017022267.1 XP_016877756.1 7 - 8 833 61
XM_017022268.1 XP_016877757.1 7 - 8 776 61
XM_017022269.1 XP_016877758.1 7 - 8 826 61
XM_017022270.1 XP_016877759.1 8 - 9 984 61
XM_017022271.1 XP_016877760.1 7 - 8 834 61
XM_017022272.1 XP_016877761.1 7 - 8 828 61
XM_017022273.1 XP_016877762.1 6 - 7 708 61
XM_017022274.1 XP_016877763.1 7 - 8 833 61
XM_017022276.1 XP_016877765.1 4 - 5 481 61
XM_017022279.1 XP_016877768.1 7 - 8 830 61
XM_017022280.1 XP_016877769.1 7 - 8 830 61
XM_017022281.1 XP_016877770.1 7 - 8 830 61
XM_017022282.1 XP_016877771.1 7 - 8 829 61
XM_017022283.1 XP_016877772.1 7 - 8 828 61
XM_017022284.1 XP_016877773.1 7 - 8 841 61
GenBank mRNA AB040887.1 - 6 - 7 658 61
AF242528.1 - 6 - 7 646 61
AK056095.1 - 7 - 8 817 61
AK056515.1 - 9 - 10 955 61
BC070197.1 - 7 - 8 688 61
BC107415.1 - 7 - 8 712 61
BX647285.1 - 6 - 7 655 61

View Full Product Details