Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 9 - 10 1238 83
NM_020843.2 NP_065894.2 8 - 9 1095 83
XM_005254417.2 XP_005254474.1 9 - 10 1252 83
XM_005254419.2 XP_005254476.1 9 - 10 1252 83
XM_011521650.1 XP_011519952.1 10 - 11 1337 83
XM_011521652.2 XP_011519954.1 8 - 9 1048 83
XM_011521653.2 XP_011519955.1 8 - 9 1132 83
XM_011521654.2 XP_011519956.1 2 - 3 396 83
XM_017022266.1 XP_016877755.1 9 - 10 1257 83
XM_017022267.1 XP_016877756.1 9 - 10 1257 83
XM_017022268.1 XP_016877757.1 9 - 10 1200 83
XM_017022269.1 XP_016877758.1 9 - 10 1250 83
XM_017022270.1 XP_016877759.1 10 - 11 1408 83
XM_017022271.1 XP_016877760.1 9 - 10 1258 83
XM_017022272.1 XP_016877761.1 9 - 10 1252 83
XM_017022273.1 XP_016877762.1 8 - 9 1132 83
XM_017022274.1 XP_016877763.1 9 - 10 1257 83
XM_017022275.1 XP_016877764.1 8 - 9 1089 83
XM_017022276.1 XP_016877765.1 6 - 7 944 83
XM_017022277.1 XP_016877766.1 8 - 9 1089 83
XM_017022278.1 XP_016877767.1 7 - 8 971 83
XM_017022279.1 XP_016877768.1 9 - 10 1254 83
XM_017022280.1 XP_016877769.1 9 - 10 1254 83
XM_017022281.1 XP_016877770.1 9 - 10 1254 83
XM_017022282.1 XP_016877771.1 9 - 10 1253 83
XM_017022283.1 XP_016877772.1 9 - 10 1252 83
XM_017022284.1 XP_016877773.1 9 - 10 1265 83
GenBank mRNA AB040887.1 - 8 - 9 1082 83
AF119814.1 - 2 - 3 302 83
AF242528.1 - 8 - 9 1070 83
AK056095.1 - 9 - 10 1241 83
AK056515.1 - 11 - 12 1379 83
BC070197.1 - 9 - 10 1112 83
BC107415.1 - 9 - 10 1136 83
BX647285.1 - 9 - 10 1238 83

View Full Product Details