The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005334.2 | NP_005325.2 | 11 - 12 | 2557 | 70 |
XM_006724815.2 | XP_006724878.1 | 11 - 12 | 3022 | 70 | |
XM_006724816.2 | XP_006724879.1 | 11 - 12 | 3022 | 70 | |
XM_011531147.2 | XP_011529449.1 | 11 - 12 | 3022 | 70 | |
XM_011531148.2 | XP_011529450.1 | 11 - 12 | 3022 | 70 | |
XM_017029471.1 | XP_016884960.1 | 10 - 11 | 2824 | 70 | |
XM_017029472.1 | XP_016884961.1 | 6 - 7 | 1210 | 70 | |
GenBank mRNA | L20010.1 | - | 11 - 12 | 2373 | 70 |