The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005274651.2 | XP_005274708.1 | 1 - 2 | 48 | 66 |
XM_017029280.1 | XP_016884769.1 | 1 - 2 | 48 | 66 | |
GenBank mRNA | AK296067.1 | - | 1 - 2 | 41 | 66 |