The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000226.3 | NP_000217.2 | 6 - 7 | 79 | |
GenBank mRNA | BC121170.2 | - | 7 - 8 | 930 | 79 |
S69510.1 | - | 6 - 7 | 79 | ||
Z29074.1 | - | 6 - 7 | 79 |