Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
Chromosome Location
Chr.6: 31639028 - 31660900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098534.1 NP_001092004.1 16 - 17 2562 57
NM_001199697.1 NP_001186626.1 16 - 17 2314 57
NM_001199698.1 NP_001186627.1 16 - 17 2692 57
NM_004639.3 NP_004630.3 16 - 17 2734 57
NM_080702.2 NP_542433.1 16 - 17 2692 57
NM_080703.2 NP_542434.1 16 - 17 2716 57
XM_011514892.2 XP_011513194.1 17 - 18 3816 57
XM_017011275.1 XP_016866764.1 17 - 18 3843 57
XM_017011276.1 XP_016866765.1 17 - 18 2833 57
XM_017011277.1 XP_016866766.1 17 - 18 2693 57
XM_017011278.1 XP_016866767.1 17 - 18 2768 57
XM_017011279.1 XP_016866768.1 17 - 18 3163 57
XM_017011280.1 XP_016866769.1 17 - 18 2735 57
XM_017011281.1 XP_016866770.1 17 - 18 2897 57
XM_017011282.1 XP_016866771.1 17 - 18 2689 57
XM_017011283.1 XP_016866772.1 17 - 18 3840 57
XM_017011284.1 XP_016866773.1 17 - 18 3810 57
XM_017011285.1 XP_016866774.1 16 - 17 2633 57
XM_017011286.1 XP_016866775.1 16 - 17 2652 57
XM_017011287.1 XP_016866776.1 17 - 18 3783 57
XM_017011288.1 XP_016866777.1 16 - 17 2570 57
XM_017011289.1 XP_016866778.1 16 - 17 3702 57
XM_017011290.1 XP_016866779.1 17 - 18 3843 57
XM_017011291.1 XP_016866780.1 17 - 18 3843 57
XM_017011292.1 XP_016866781.1 16 - 17 3672 57
XM_017011293.1 XP_016866782.1 15 - 16 2465 57
XM_017011294.1 XP_016866783.1 15 - 16 3597 57
XM_017011295.1 XP_016866784.1 17 - 18 3843 57
XM_017011296.1 XP_016866785.1 16 - 17 3672 57
XM_017011297.1 XP_016866786.1 16 - 17 2570 57
XM_017011298.1 XP_016866787.1 16 - 17 3672 57
GenBank mRNA AK302695.1 - 16 - 17 2265 57
AK304879.1 - 16 - 17 2643 57
BC003133.1 - 16 - 17 2610 57
BX647244.1 - 16 - 17 2570 57
M33519.1 - 16 - 17 2669 57

View Full Product Details