The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098534.1 | NP_001092004.1 | 1 - 2 | 145 | 82 |
XM_017011277.1 | XP_016866766.1 | 1 - 2 | 108 | 82 | |
XM_017011286.1 | XP_016866775.1 | 1 - 2 | 121 | 82 | |
GenBank mRNA | BX647244.1 | - | 1 - 2 | 153 | 82 |
CX758454.1 | - | 1 - 2 | 120 | 82 |