The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000817.2 | NP_000808.2 | 4 - 5 | 732 | 73 |
NM_013445.3 | NP_038473.2 | 4 - 5 | 736 | 73 | |
XM_005246444.2 | XP_005246501.1 | 4 - 5 | 730 | 73 | |
XM_011510922.1 | XP_011509224.1 | 4 - 5 | 586 | 73 | |
XM_017003756.1 | XP_016859245.1 | 4 - 5 | 535 | 73 | |
XM_017003757.1 | XP_016859246.1 | 4 - 5 | 730 | 73 | |
XM_017003758.1 | XP_016859247.1 | 4 - 5 | 730 | 73 | |
GenBank mRNA | A28074.1 | - | 3 - 4 | 310 | 73 |
AF178853.2 | - | 3 - 4 | 310 | 73 | |
AK292582.1 | - | 4 - 5 | 535 | 73 | |
BC002815.2 | - | 4 - 5 | 514 | 73 | |
BC026349.1 | - | 4 - 5 | 536 | 73 | |
BC036552.1 | - | 3 - 4 | 1139 | 73 | |
BC037780.2 | - | 4 - 5 | 732 | 73 | |
KU178086.1 | - | 3 - 4 | 310 | 73 | |
L16888.1 | - | 4 - 5 | 493 | 73 | |
M81883.1 | - | 4 - 5 | 860 | 73 | |
S61897.1 | - | 3 - 4 | 319 | 73 | |
S61898.1 | - | 4 - 5 | 514 | 73 | |
Z22750.1 | - | 4 - 5 | 493 | 73 |