The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320350.1 | NP_001307279.1 | 11 - 12 | 1895 | 105 |
NM_001320351.1 | NP_001307280.1 | 11 - 12 | 1895 | 105 | |
NM_003307.3 | NP_003298.1 | 11 - 12 | 1895 | 105 | |
NR_038257.1 | 11 - 12 | 1895 | 105 | ||
XM_005261171.3 | XP_005261228.1 | 12 - 13 | 2056 | 105 | |
XM_011529736.2 | XP_011528038.1 | 11 - 12 | 2299 | 105 | |
XM_017028456.1 | XP_016883945.1 | 12 - 13 | 2059 | 105 | |
XM_017028457.1 | XP_016883946.1 | 11 - 12 | 2300 | 105 | |
GenBank mRNA | AB001535.1 | - | 11 - 12 | 2240 | 105 |
AB166745.1 | - | 12 - 13 | 2008 | 105 | |
AB166746.1 | - | 7 - 8 | 1247 | 105 | |
AJ417076.1 | - | 12 - 13 | 1748 | 105 | |
AJ878416.1 | - | 11 - 12 | 1858 | 105 | |
AY603182.1 | - | 11 - 12 | 1795 | 105 | |
BC112342.1 | - | 11 - 12 | 1808 | 105 | |
BI755424.1 | - | 1 - 2 | 253 | 105 | |
BI906484.1 | - | 4 - 5 | 660 | 106 |