Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM2
Entrez Gene ID
7226
Gene Name
transient receptor potential cation channel subfamily M member 2
Gene Aliases
EREG1, KNP3, LTRPC2, LTrpC-2, NUDT9H, NUDT9L1, TRPC7
Chromosome Location
Chr.21: 44350112 - 44443081 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320350.1 NP_001307279.1 29 - 30 4312 97
NM_001320351.1 NP_001307280.1 27 - 28 4060 97
NM_001320352.1 NP_001307281.1 3 - 4 424 97
NM_003307.3 NP_003298.1 28 - 29 4162 97
NR_038257.1 28 - 29 4165 97
XM_005261171.3 XP_005261228.1 29 - 30 4323 97
XM_011529736.2 XP_011528038.1 29 - 30 4656 97
XM_017028456.1 XP_016883945.1 30 - 31 4416 97
XM_017028457.1 XP_016883946.1 28 - 29 4555 97
GenBank mRNA AB001535.1 - 28 - 29 4507 97
AB017549.1 - 4 - 5 393 97
AB166745.1 - 29 - 30 4275 97
AB166746.1 - 24 - 25 3514 97
AJ417076.1 - 28 - 29 3913 97
AJ878416.1 - 28 - 29 4125 97
AK301101.1 - 3 - 4 276 97
AK301451.1 - 9 - 10 1115 97
BC112342.1 - 29 - 30 4225 97
EU362987.1 - 3 - 4 424 97

View Full Product Details