The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320350.1 | NP_001307279.1 | 30 - 31 | 4417 | 82 |
NM_001320351.1 | NP_001307280.1 | 28 - 29 | 4165 | 82 | |
NM_001320352.1 | NP_001307281.1 | 4 - 5 | 529 | 82 | |
NM_003307.3 | NP_003298.1 | 29 - 30 | 4267 | 82 | |
NR_038257.1 | 29 - 30 | 4270 | 82 | ||
XM_005261171.3 | XP_005261228.1 | 30 - 31 | 4428 | 82 | |
XM_011529736.2 | XP_011528038.1 | 30 - 31 | 4761 | 82 | |
XM_017028456.1 | XP_016883945.1 | 31 - 32 | 4521 | 82 | |
XM_017028457.1 | XP_016883946.1 | 29 - 30 | 4660 | 82 | |
GenBank mRNA | AB001535.1 | - | 29 - 30 | 4612 | 82 |
AB017549.1 | - | 5 - 6 | 498 | 82 | |
AB166745.1 | - | 30 - 31 | 4380 | 82 | |
AB166746.1 | - | 25 - 26 | 3619 | 82 | |
AJ417076.1 | - | 29 - 30 | 4018 | 82 | |
AJ878416.1 | - | 29 - 30 | 4230 | 82 | |
AK301101.1 | - | 4 - 5 | 381 | 82 | |
AK301451.1 | - | 10 - 11 | 1220 | 82 | |
BC112342.1 | - | 30 - 31 | 4330 | 82 | |
EU362987.1 | - | 4 - 5 | 529 | 82 |