The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320350.1 | NP_001307279.1 | 4 - 5 | 707 | 87 |
NM_001320351.1 | NP_001307280.1 | 4 - 5 | 707 | 87 | |
NM_003307.3 | NP_003298.1 | 4 - 5 | 707 | 87 | |
NR_038257.1 | 4 - 5 | 707 | 87 | ||
XM_005261171.3 | XP_005261228.1 | 5 - 6 | 868 | 87 | |
XM_011529736.2 | XP_011528038.1 | 4 - 5 | 1111 | 87 | |
XM_017028456.1 | XP_016883945.1 | 5 - 6 | 871 | 87 | |
XM_017028457.1 | XP_016883946.1 | 4 - 5 | 1112 | 87 | |
GenBank mRNA | AB001535.1 | - | 4 - 5 | 1052 | 87 |
AB166745.1 | - | 5 - 6 | 820 | 87 | |
AJ417076.1 | - | 4 - 5 | 620 | 87 | |
AJ878416.1 | - | 4 - 5 | 670 | 87 | |
AY603182.1 | - | 4 - 5 | 607 | 87 | |
BC112342.1 | - | 4 - 5 | 620 | 87 |