Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SON
Entrez Gene ID
6651
Gene Name
SON DNA binding protein
Gene Aliases
BASS1, C21orf50, DBP-5, NREBP, SON3
Chromosome Location
Chr.21: 33543038 - 33577514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103797.1 8 - 9 6941 78
GenBank mRNA AF380181.1 - 8 - 9 6915 78

View Full Product Details