Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location
Chr.8: 140658382 - 141001313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199649.1 NP_001186578.1 26 - 27 2805 91
NM_001316342.1 NP_001303271.1 26 - 27 2588 91
NM_005607.4 NP_005598.3 26 - 27 2794 91
NM_153831.3 NP_722560.1 26 - 27 2805 91
XM_005251003.2 XP_005251060.1 28 - 29 2969 91
XM_005251012.2 XP_005251069.1 28 - 29 2811 91
XM_005251013.2 XP_005251070.1 27 - 28 2630 91
XM_006716608.2 XP_006716671.1 29 - 30 2900 91
XM_006716609.2 XP_006716672.1 28 - 29 2733 91
XM_006716610.2 XP_006716673.1 28 - 29 2725 91
XM_011517194.2 XP_011515496.1 29 - 30 2869 91
XM_017013651.1 XP_016869140.1 27 - 28 2650 91
XM_017013653.1 XP_016869142.1 27 - 28 2885 91
XM_017013654.1 XP_016869143.1 26 - 27 2867 91
XM_017013655.1 XP_016869144.1 27 - 28 2861 91
XM_017013656.1 XP_016869145.1 26 - 27 2867 91
XM_017013657.1 XP_016869146.1 25 - 26 2846 91
XM_017013658.1 XP_016869147.1 28 - 29 2772 91
XM_017013660.1 XP_016869149.1 28 - 29 2709 91
XM_017013661.1 XP_016869150.1 27 - 28 2727 91
XM_017013662.1 XP_016869151.1 27 - 28 2691 91
XM_017013663.1 XP_016869152.1 26 - 27 2622 91
XM_017013664.1 XP_016869153.1 27 - 28 2795 91
XM_017013665.1 XP_016869154.1 26 - 27 2619 91
XM_017013666.1 XP_016869155.1 24 - 25 2738 91
XM_017013667.1 XP_016869156.1 28 - 29 2685 91
XM_017013668.1 XP_016869157.1 27 - 28 2703 91
XM_017013669.1 XP_016869158.1 27 - 28 2691 91
XM_017013670.1 XP_016869159.1 27 - 28 2688 91
XM_017013671.1 XP_016869160.1 26 - 27 2670 91
XM_017013672.1 XP_016869161.1 25 - 26 2601 91
XM_017013673.1 XP_016869162.1 25 - 26 2601 91
XM_017013677.1 XP_016869166.1 27 - 28 2601 91
XM_017013678.1 XP_016869167.1 24 - 25 2493 91
XM_017013680.1 XP_016869169.1 28 - 29 3785 91
XM_017013681.1 XP_016869170.1 28 - 29 2733 91
XM_017013682.1 XP_016869171.1 26 - 27 2609 91
XM_017013683.1 XP_016869172.1 27 - 28 2649 91
XM_017013684.1 XP_016869173.1 26 - 27 2631 91
XM_017013685.1 XP_016869174.1 26 - 27 2572 91
XM_017013686.1 XP_016869175.1 24 - 25 2507 91
XM_017013688.1 XP_016869177.1 26 - 27 2541 91
XM_017013689.1 XP_016869178.1 25 - 26 2406 91
XM_017013690.1 XP_016869179.1 24 - 25 2362 91
GenBank mRNA AB209071.1 - 26 - 27 2494 91
AB209083.1 - 29 - 30 2515 91
AK094999.1 - 16 - 17 1511 91
AK294873.1 - 17 - 18 1589 91
AK301557.1 - 5 - 6 510 91
AK304356.1 - 26 - 27 2650 91
AL832961.1 - 26 - 27 2677 91
BC028733.2 - 12 - 13 1453 91
BC035404.2 - 23 - 24 2388 91
KF437463.1 - 24 - 25 2498 91
L13616.1 - 26 - 27 2727 91

View Full Product Details