The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006724100.3 | XP_006724163.1 | 1 - 2 | 288 | 85 |
XM_006724101.3 | XP_006724164.1 | 1 - 2 | 288 | 85 | |
XM_006724102.1 | XP_006724165.1 | 1 - 2 | 77 | 85 | |
GenBank mRNA | BC017326.2 | - | 1 - 2 | 46 | 85 |
DW452042.1 | - | 1 - 2 | 127 | 85 |