The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003681.2 | NP_001003681.1 | 10 - 11 | 1937 | 67 |
NR_027780.1 | 11 - 12 | 2052 | 67 | ||
XM_006724100.3 | XP_006724163.1 | 9 - 10 | 2016 | 67 | |
XM_006724102.1 | XP_006724165.1 | 10 - 11 | 1920 | 67 | |
GenBank mRNA | AJ010069.1 | - | 5 - 6 | 548 | 67 |
AK299833.1 | - | 11 - 12 | 2017 | 67 | |
AL079310.1 | - | 11 - 12 | 2001 | 67 | |
BC104754.1 | - | 6 - 7 | 1499 | 67 | |
BC104756.1 | - | 6 - 7 | 1499 | 67 | |
CR456504.1 | - | 9 - 10 | 1793 | 67 |