Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3A1
Entrez Gene ID
10291
Gene Name
splicing factor 3a subunit 1
Gene Aliases
PRP21, PRPF21, SAP114, SF3A120
Chromosome Location
Chr.22: 30331988 - 30356947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005877.5 NP_005868.1 1 - 2 219 118
GenBank mRNA AK303273.1 - 1 - 2 194 118
BC001976.1 - 1 - 2 159 118
BC007684.2 - 1 - 2 186 118
CR456575.1 - 1 - 2 162 118
X85237.1 - 1 - 2 161 118

View Full Product Details