Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3A1
Entrez Gene ID
10291
Gene Name
splicing factor 3a subunit 1
Gene Aliases
PRP21, PRPF21, SAP114, SF3A120
Chromosome Location
Chr.22: 30331988 - 30356947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005877.5 NP_005868.1 7 - 8 1222 66
GenBank mRNA AK303273.1 - 8 - 9 1147 66
BC001976.1 - 7 - 8 1162 66
BC007684.2 - 7 - 8 1189 66
CR456575.1 - 7 - 8 1165 66
X85237.1 - 7 - 8 1164 66

View Full Product Details