The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006377.3 | NP_006368.3 | 32 - 33 | 4120 | 59 |
XM_011517681.2 | XP_011515983.1 | 25 - 26 | 9284 | 59 | |
XM_011517682.2 | XP_011515984.1 | 25 - 26 | 9282 | 59 | |
XM_011517683.2 | XP_011515985.1 | 25 - 26 | 9280 | 59 | |
XM_011517684.2 | XP_011515986.1 | 32 - 33 | 4138 | 59 | |
XM_011517685.2 | XP_011515987.1 | 32 - 33 | 4135 | 59 | |
XM_011517686.2 | XP_011515988.1 | 23 - 24 | 3312 | 59 | |
XM_017014191.1 | XP_016869680.1 | 23 - 24 | 3173 | 59 | |
XM_017014192.1 | XP_016869681.1 | 25 - 26 | 3090 | 59 | |
GenBank mRNA | AB209998.1 | - | 32 - 33 | 4103 | 59 |
AF020202.1 | - | 32 - 33 | 4052 | 59 | |
BC111781.1 | - | 32 - 33 | 4008 | 59 | |
BC111800.1 | - | 34 - 35 | 4037 | 59 |