The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206774.1 | NP_001193703.1 | 4 - 5 | 689 | 71 |
NM_012086.4 | NP_036218.1 | 4 - 5 | 689 | 71 | |
XM_005246965.4 | XP_005247022.1 | 4 - 5 | 604 | 71 | |
GenBank mRNA | AF133123.1 | - | 4 - 5 | 625 | 71 |
AF465407.1 | - | 4 - 5 | 580 | 71 | |
AK025560.1 | - | 4 - 5 | 603 | 71 | |
AK314718.1 | - | 4 - 5 | 630 | 71 | |
BC043347.1 | - | 4 - 5 | 544 | 71 | |
S81493.1 | - | 1 - 2 | 52 | 71 |