Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALS2
Entrez Gene ID
57679
Gene Name
ALS2, alsin Rho guanine nucleotide exchange factor
Gene Aliases
ALS2CR6, ALSJ, IAHSP, PLSJ
Chromosome Location
Chr.2: 201700263 - 201781172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020919.3 NP_065970.2 4 - 5 1470 72
XM_006712654.2 XP_006712717.1 4 - 5 2047 72
XM_011511530.2 XP_011509832.1 3 - 4 1182 72
XM_017004569.1 XP_016860058.1 3 - 4 1174 72
XM_017004570.1 XP_016860059.1 3 - 4 1174 72
XM_017004571.1 XP_016860060.1 3 - 4 1174 72
XM_017004572.1 XP_016860061.1 1 - 2 659 72
GenBank mRNA AB046783.2 - 3 - 4 72
AB053305.1 - 4 - 5 72
AF391100.1 - 4 - 5 1237 72
AK291029.1 - 4 - 5 1254 72
BC029174.1 - 4 - 5 72
BC071576.1 - 4 - 5 1232 72
KU178735.1 - 3 - 4 1114 72
KU178736.1 - 3 - 4 1114 72

View Full Product Details