The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033066.2 | NP_149055.1 | 16 - 17 | 1335 | 64 |
XM_017004620.1 | XP_016860109.1 | 12 - 13 | 2811 | 64 | |
GenBank mRNA | AB053302.1 | - | 12 - 13 | 1203 | 64 |
AB053304.1 | - | 7 - 8 | 1405 | 64 | |
AF316032.1 | - | 16 - 17 | 1335 | 64 | |
AK131208.1 | - | 13 - 14 | 1248 | 64 | |
AK298589.1 | - | 11 - 12 | 1190 | 64 | |
AK298947.1 | - | 13 - 14 | 1220 | 64 | |
AK300622.1 | - | 11 - 12 | 1128 | 64 | |
BC132785.1 | - | 14 - 15 | 1211 | 64 | |
BC144206.1 | - | 11 - 12 | 1139 | 64 | |
BX647162.1 | - | 19 - 20 | 1506 | 64 |