The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033066.2 | NP_149055.1 | 3 - 4 | 402 | 108 |
XM_017004620.1 | XP_016860109.1 | 3 - 4 | 1971 | 108 | |
GenBank mRNA | AB053302.1 | - | 3 - 4 | 402 | 108 |
AB053303.1 | - | 3 - 4 | 339 | 108 | |
AF316032.1 | - | 3 - 4 | 402 | 108 | |
AK131208.1 | - | 3 - 4 | 420 | 108 | |
AK298589.1 | - | 3 - 4 | 431 | 108 | |
AK298947.1 | - | 3 - 4 | 359 | 108 | |
AK300622.1 | - | 3 - 4 | 420 | 108 | |
BC132785.1 | - | 2 - 3 | 299 | 108 | |
BC144206.1 | - | 2 - 3 | 299 | 108 | |
BX647162.1 | - | 3 - 4 | 294 | 108 | |
BX648531.1 | - | 3 - 4 | 308 | 108 |