Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM8
Entrez Gene ID
79054
Gene Name
transient receptor potential cation channel subfamily M member 8
Gene Aliases
LTRPC6, TRPP8
Chromosome Location
Chr.2: 233917342 - 234019522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024080.4 NP_076985.4 9 - 10 1182 94
XM_011511810.2 XP_011510112.1 9 - 10 1238 94
XM_017004891.1 XP_016860380.1 8 - 9 3049 94
GenBank mRNA AB061779.1 - 8 - 9 1115 94
AY090109.1 - 9 - 10 1182 94
AY328400.1 - 8 - 9 1142 94
BC126342.1 - 10 - 11 1432 94
BC143819.1 - 10 - 11 1432 94
DQ139309.1 - 8 - 9 1142 94
KC692993.1 - 9 - 10 1209 94

View Full Product Details