The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014169.3 | NP_054888.2 | 5 - 6 | 1000 | 92 |
GenBank mRNA | AB100262.2 | - | 5 - 6 | 635 | 92 |
AF161483.1 | - | 5 - 6 | 657 | 92 | |
AF212243.1 | - | 5 - 6 | 630 | 92 | |
AW732226.1 | - | 1 - 2 | 92 | ||
AY329084.1 | - | 5 - 6 | 609 | 92 | |
BC010893.1 | - | 5 - 6 | 637 | 92 | |
BC107699.1 | - | 5 - 6 | 662 | 92 | |
BC113533.1 | - | 5 - 6 | 1000 | 92 | |
BC113535.1 | - | 5 - 6 | 1000 | 92 | |
CB241867.1 | - | 1 - 2 | 92 |