The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144920.2 | NP_001138392.1 | 3 - 4 | 346 | 108 |
NM_001287512.1 | NP_001274441.1 | 3 - 4 | 346 | 108 | |
NM_001287513.1 | NP_001274442.1 | 3 - 4 | 275 | 108 | |
NM_001318333.1 | NP_001305262.1 | 3 - 4 | 346 | 108 | |
NM_001321740.1 | NP_001308669.1 | 3 - 4 | 346 | 108 | |
NM_001321741.1 | NP_001308670.1 | 3 - 4 | 346 | 108 | |
NM_001321742.1 | NP_001308671.1 | 3 - 4 | 346 | 108 | |
NM_024926.3 | NP_079202.2 | 3 - 4 | 346 | 108 | |
GenBank mRNA | AK022633.1 | - | 3 - 4 | 312 | 108 |
AK293444.1 | - | 3 - 4 | 197 | 108 | |
AK295092.1 | - | 3 - 4 | 326 | 108 | |
AK300807.1 | - | 3 - 4 | 312 | 108 | |
AL137393.1 | - | 3 - 4 | 310 | 108 | |
BC013912.1 | - | 3 - 4 | 315 | 108 | |
BC126331.1 | - | 3 - 4 | 271 | 108 | |
BC130339.1 | - | 3 - 4 | 271 | 108 | |
BP315162.1 | - | 3 - 4 | 346 | 108 |