Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC26
Entrez Gene ID
79989
Gene Name
tetratricopeptide repeat domain 26
Gene Aliases
DYF13, IFT56, dyf-13
Chromosome Location
Chr.7: 139133744 - 139191986 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144920.2 NP_001138392.1 9 - 10 1001 99
NM_001144923.2 NP_001138395.1 8 - 9 908 99
NM_001287512.1 NP_001274441.1 6 - 7 608 99
NM_001287513.1 NP_001274442.1 9 - 10 930 99
NM_001318333.1 NP_001305262.1 9 - 10 1162 99
NM_001321740.1 NP_001308669.1 9 - 10 1001 99
NM_001321741.1 NP_001308670.1 7 - 8 683 99
NM_024926.3 NP_079202.2 9 - 10 1001 99
GenBank mRNA AK022633.1 - 9 - 10 967 99
AK293444.1 - 9 - 10 852 99
AK295092.1 - 6 - 7 588 99
AK299132.1 - 8 - 9 869 99
AL137393.1 - 9 - 10 965 99
BC126331.1 - 9 - 10 926 99
BC130339.1 - 9 - 10 926 99

View Full Product Details